Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162946.1 | NP_001156418.1 | 20 | 3380 | ||
NM_008797.3 | NP_032823.2 | 19 | 3357 | |||
GenBank mRNA | AK077665.1 | - | 11 | 2185 | ||
AK128927.1 | - | 3154 | ||||
AK148328.1 | - | 20 | 3327 | |||
AK170596.1 | - | 20 | 3340 | |||
AK170871.1 | - | 20 | 3331 | |||
AK172320.1 | - | 20 | 3308 | |||
BC055030.1 | - | 20 | 3315 | 5053475 | ||
L09192.1 | - | 19 | 3333 | |||
M97957.1 | - | 14 | 2505 |