Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008894.2 | NP_032920.2 | 4 | 566 | ||
GenBank mRNA | AK013901.1 | - | 4 | 533 | ||
AK218431.1 | - | 3 | 281 | |||
BC003342.1 | - | 4 | 532 | 3257610 | ||
BG078511.2 | - | 4 | 519 | |||
CB205181.1 | - | 1 | 102 | |||
Z72486.1 | - | 4 | 527 |