Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008997.3 | NP_033023.1 | 2 | 415 | ||
GenBank mRNA | AB232606.1 | - | 2 | 191 | ||
AK134125.1 | - | 2 | 160 | |||
AK146452.1 | - | 2 | 276 | |||
AK146460.1 | - | 2 | 227 | |||
AK146623.1 | - | 2 | 227 | |||
AK146978.1 | - | 2 | 207 | |||
AK147077.1 | - | 2 | 246 | |||
AK148022.1 | - | 2 | 288 | |||
AK148106.1 | - | 2 | 214 | |||
AK177220.1 | - | 2 | 132 | |||
AK186595.1 | - | 2 | 182 | |||
AK189626.1 | - | 2 | 182 | |||
AK195913.1 | - | 1 | 71 | |||
BC054753.1 | - | 2 | 231 | 6411651 | ||
BC085270.1 | - | 2 | 224 | 6478784 | ||
BC157916.1 | - | 2 | 244 | 9007344 | ||
BU705914.1 | - | 2 | 415 | |||
BY275360.1 | - | 2 | 410 | |||
KJ650201.1 | - | 2 | 191 | |||
L26528.1 | - | 2 | 226 |