Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163557.1 | NP_001157029.1 | 4 | 419 | ||
NM_008905.2 | NP_032931.2 | 4 | 655 | |||
GenBank mRNA | AK035364.1 | - | 4 | 423 | ||
AK135964.1 | - | 4 | 690 | |||
BB627669.1 | - | 4 | 420 | |||
BC051202.1 | - | 4 | 503 | |||
BC086931.1 | - | 4 | 774 | |||
BC100346.1 | - | 4 | 419 | 30936010 | ||
U79024.1 | - | 4 | 440 |