Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008966.3 | NP_032992.1 | 2 | 496 | ||
GenBank mRNA | AB094411.1 | - | 1 | 197 | ||
AK014673.1 | - | 1 | 352 | |||
AK076450.1 | - | 2 | 498 | |||
AK087833.1 | - | 2 | 401 | |||
AK141760.1 | - | 2 | 471 | |||
BC064794.1 | - | 2 | 439 | 30429190 | ||
D17433.1 | - | 2 | 420 |
Set Membership: |
G-protein coupled receptor |