Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou3f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008899.2 | NP_032925.1 | 1 | 1463 | ||
GenBank mRNA | X66602.1 | - | 1 | 1395 |