Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008997.3 | NP_033023.1 | 5 | 1356 | ||
GenBank mRNA | AK134125.1 | - | 5 | 1101 | ||
AK146452.1 | - | 5 | 1217 | |||
AK146460.1 | - | 5 | 1168 | |||
AK146623.1 | - | 5 | 1168 | |||
AK146978.1 | - | 5 | 1148 | |||
AK147077.1 | - | 5 | 1187 | |||
AK148022.1 | - | 5 | 1230 | |||
AK185489.1 | - | 1 | 197 | |||
BC054753.1 | - | 5 | 1172 | 6411651 | ||
BC085270.1 | - | 5 | 1166 | 6478784 | ||
L26528.1 | - | 5 | 1168 |