Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039966.1 | NP_001035055.1 | 3 | 1306 | ||
NM_001098201.1 | NP_001091671.1 | 2 | 979 | |||
NM_001505.2 | NP_001496.1 | 2 | 1182 | |||
GenBank mRNA | AB451427.1 | - | 1 | 422 | ||
AF015257.1 | - | 2 | 1113 | |||
AK289588.1 | - | 2 | 953 | |||
BC011634.1 | - | 1 | 513 | 4079700 | ||
BC082766.1 | - | 1 | 523 | 5419168 | ||
CR541655.1 | - | 1 | 422 | |||
CR541741.1 | - | 1 | 422 | |||
U63917.1 | - | 2 | 926 | |||
X98510.1 | - | 1 | 595 | |||
Y08162.1 | - | 1 | 422 |
Set Membership: |
![]() |