Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009068.3 | NP_033094.3 | 3 | 400 | ||
GenBank mRNA | AK030959.1 | - | 3 | 631 | ||
AK048989.1 | - | 3 | 628 | |||
AK156803.1 | - | 3 | 665 | |||
BC050905.1 | - | 3 | 400 | 5721177 | ||
BC054542.1 | - | 3 | 400 | 5721177 | ||
BC058162.1 | - | 3 | 586 | 5135235 | ||
U25995.1 | - | 2 | 315 |