Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009093.2 | NP_033119.1 | 3 | 308 | ||
GenBank mRNA | AA217935.1 | - | 3 | 192 | ||
AK002939.1 | - | 3 | 212 | |||
AK012285.1 | - | 3 | 211 | |||
AK224925.1 | - | 3 | 201 | |||
BC024393.1 | - | 228 | 4986748 | |||
BC051203.1 | - | 3 | 195 | 1476903 | ||
BC055750.1 | - | 3 | 259 | |||
L31609.1 | - | 3 | 192 |