Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088415.1 | - | 7 | 1194 | ||
AK088805.1 | - | 7 | 1182 | |||
AK144136.1 | - | 7 | 1189 | |||
BC005455.1 | - | 7 | 1161 | 3495818 | ||
X53247.1 | - | 7 | 1141 |