Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166175.1 | NP_001159647.1 | 1 | 549 | ||
NM_001166176.1 | NP_001159648.1 | 1 | 549 | |||
NM_004387.3 | NP_004378.1 | 1 | 549 | |||
XM_017009071.1 | XP_016864560.1 | 1 | 443 | |||
GenBank mRNA | AB021133.1 | - | 1 | 320 | ||
AK290615.1 | - | 1 | 441 | |||
AK297844.1 | - | 1 | 443 | |||
AK307218.1 | - | 1 | 443 | |||
AK307247.1 | - | 1 | 501 | |||
AK307249.1 | - | 1 | 442 | |||
AK309495.1 | - | 1 | 443 | |||
BC025711.1 | - | 1 | 427 | 5225103 | ||
DA559427.1 | - | 1 | 549 | |||
KU178001.1 | - | 1 | 320 | |||
KU178002.1 | - | 1 | 320 | |||
U34962.1 | - | 1 | 496 |