Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009138.3 | NP_033164.1 | 2, 3 | 199 | ||
GenBank mRNA | AJ249480.1 | - | 2, 3 | 172 | ||
AK131875.1 | - | 2, 3 | 200 | |||
AK134138.1 | - | 2, 3 | 184 | |||
BC117033.1 | - | 1, 2 | 132 | 40126352 | ||
BC117037.1 | - | 1, 2 | 132 | 40126356 | ||
DQ158256.1 | - | 2, 3 | 185 | |||
U86357.1 | - | 1, 2 | 81 |