Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009156.2 | NP_033182.1 | 3 | 168 | ||
GenBank mRNA | AF015284.1 | - | 3 | 123 | ||
AF241527.2 | - | 3 | 121 | |||
AK002870.1 | - | 3 | 144 | |||
AK028124.1 | - | 3 | 138 | |||
BC052719.1 | - | 3 | 130 | 6814242 | ||
BU604713.1 | - | 3 | 201 | |||
U67890.1 | - | 3 | 120 |