Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009139.3 | NP_033165.1 | 4 | 473 | ||
GenBank mRNA | AF128188.1 | - | 4 | 369 | ||
AF128189.1 | - | 4 | 369 | |||
AF128190.1 | - | 4 | 369 | |||
AF128191.1 | - | 4 | 369 | |||
AF128192.1 | - | 4 | 369 | |||
AK002697.1 | - | 4 | 376 | |||
AK008547.1 | - | 4 | 377 | |||
AK150336.1 | - | 4 | 376 | |||
AK154553.1 | - | 4 | 377 | |||
M58004.1 | - | 4 | 376 |