Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009144.2 | NP_033170.1 | 3 | 832 | ||
GenBank mRNA | AF017989.1 | - | 3 | 864 | ||
AK138252.1 | - | 3 | 671 | |||
AK147091.1 | - | 3 | 829 | |||
BC014722.1 | - | 3 | 792 | 4487469 | ||
D50462.1 | - | 3 | 624 | |||
U88567.1 | - | 3 | 846 |
Set Membership: |
![]() |