Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159523.1 | NP_001152995.1 | 4 | 794 | ||
NM_009168.2 | NP_033194.1 | 4 | 791 | |||
GenBank mRNA | AB018423.1 | - | 4 | 790 | ||
AK142491.1 | - | 4 | 807 | |||
AK164639.1 | - | 4 | 727 | |||
BC052026.1 | - | 3 | 1721 | |||
BC079574.1 | - | 3 | 1721 | |||
BC103798.1 | - | 4 | 687 | 30288676 |