Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Soat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009230.3 | NP_033256.2 | 4 | 390 | ||
GenBank mRNA | AK054149.1 | - | 4 | 303 | ||
AK142486.1 | - | 4 | 307 | |||
AK159393.1 | - | 4 | 320 | |||
AK159529.1 | - | 4 | 320 | |||
AK160017.1 | - | 4 | 320 | |||
BC083092.1 | - | 3 | 204 | 30446741 | ||
CJ164331.1 | - | 4 | 393 | |||
DQ903181.1 | - | 4 | 307 | |||
S81092.1 | - | 4 | 324 |