Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009299.2 | NP_033325.2 | 2 | 211 | ||
GenBank mRNA | BC104304.1 | - | 2 | 209 | 40043712 | |
BC104305.1 | - | 2 | 211 | 40043713 | ||
CJ055470.1 | - | 2 | 211 | |||
M94179.1 | - | 2 | 197 |