Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009228.2 | NP_033254.2 | 2, 3 | 586 | ||
GenBank mRNA | AK137379.1 | - | 2, 3 | 587 | ||
BB625367.1 | - | 2, 3 | 579 | |||
BC018546.1 | - | 2, 3 | 536 | 3674921 | ||
U00677.1 | - | 2, 3 | 583 |