Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009292.1 | NP_033318.1 | 2 | 265 | ||
GenBank mRNA | AK006887.1 | - | 3 | 609 | ||
BC103590.1 | - | 2 | 211 | 40045824 | ||
BC103591.1 | - | 2 | 211 | 40045823 | ||
BC103603.1 | - | 2 | 208 | |||
Z75287.1 | - | 2 | 265 |