Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008405.2 | NP_001008405.1 | 2 | 172 | ||
NM_018844.3 | NP_061332.2 | 3 | 497 | |||
NR_027830.1 | 2 | 391 | ||||
XM_006716051.2 | XP_006716114.1 | 3 | 493 | |||
XM_011516406.2 | XP_011514708.1 | 3 | 256 | |||
XM_011516407.1 | XP_011514709.1 | 3 | 493 | |||
XM_011516408.2 | XP_011514710.1 | 3 | 493 | |||
XM_011516409.2 | XP_011514711.1 | 3 | 493 | |||
XM_017012436.1 | XP_016867925.1 | 3 | 493 | |||
XM_017012437.1 | XP_016867926.1 | 3 | 493 | |||
XM_017012438.1 | XP_016867927.1 | 3 | 493 | |||
GenBank mRNA | AF126020.1 | - | 3 | 204 | ||
AK289401.1 | - | 3 | 254 | |||
AK294824.1 | - | 2 | 122 | |||
BC008478.1 | - | 3 | 209 | 4294273 | ||
BG776708.1 | - | 3 | 497 | |||
BT006981.1 | - | 2 | 158 | |||
BX457671.2 | - | 3 | 212 | |||
BX647653.1 | - | 3 | 208 | |||
CK820004.1 | - | 1 | 29 |