Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286628.1 | NP_001273557.1 | 1 | 289 | ||
NM_009352.3 | NP_033378.1 | 1 | 289 | |||
GenBank mRNA | AB109563.1 | - | 1 | 269 | ||
AK021235.1 | - | 1 | 279 | |||
AK036615.1 | - | 1 | 291 | |||
AK132022.1 | - | 1 | 272 | |||
BB652245.1 | - | 1 | 290 | |||
BC034866.1 | - | 1 | 277 | 3470277 | ||
U65586.1 | - | 1 | 269 |