Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Th | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009377.1 | NP_033403.1 | 2 | 238 | ||
GenBank mRNA | M69200.1 | - | 2 | 238 |