Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009410.2 | NP_033436.1 | 1 | 366 | ||
GenBank mRNA | AB006074.1 | - | 1 | 367 | ||
AK037640.1 | - | 1 | 267 | |||
AK164443.1 | - | 1 | 267 | |||
AK170291.1 | - | 2 | 173 | |||
AK171286.1 | - | 2 | 173 | |||
BC099689.1 | - | 1 | 181 | 30731611 |