Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tbxa2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277265.1 | NP_001264194.1 | 3 | 1338 | ||
NM_009325.4 | NP_033351.1 | 4 | 1492 | |||
GenBank mRNA | AK158477.1 | - | 3 | 1311 | ||
BC089611.1 | - | 4 | 1465 | 6762421 | ||
BC138885.1 | - | 3 | 1273 | 8861907 | ||
BC138886.1 | - | 3 | 1273 | 8861908 | ||
D10849.1 | - | 3 | 1217 |
Set Membership: |
G-protein coupled receptor |