Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079822.2 | NP_001073290.1 | 4 | 590 | ||
NM_009332.3 | NP_033358.2 | 4 | 590 | |||
GenBank mRNA | AJ223069.1 | - | 4 | 498 | ||
AJ223233.1 | - | 4 | 413 | |||
AK052721.1 | - | 4 | 800 | |||
BC128305.1 | - | 4 | 497 | 40109611 | ||
BC128306.1 | - | 4 | 497 | 40109613 |