Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009398.2 | NP_033424.1 | 2, 3 | 287 | ||
GenBank mRNA | AK139985.1 | - | 2, 3 | 297 | ||
BC021155.1 | - | 2, 3 | 266 | 4208738 | ||
U83903.1 | - | 2, 3 | 287 |