Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbxa2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277265.1 | NP_001264194.1 | 3 | 1564 | ||
NM_009325.4 | NP_033351.1 | 4 | 1718 | |||
GenBank mRNA | AK158477.1 | - | 3 | 1537 | ||
BC089611.1 | - | 4 | 1691 | 6762421 | ||
D10849.1 | - | 3 | 1443 |
Set Membership: |
![]() |