Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009362.2 | NP_033388.1 | 1, 2 | 131 | ||
GenBank mRNA | AK008625.1 | - | 1, 2 | 128 | ||
AK008809.1 | - | 1, 2 | 128 | |||
AK008873.1 | - | 1, 2 | 131 | |||
AK010179.1 | - | 1, 2 | 128 | |||
AK131896.1 | - | 1, 2 | 132 | |||
AK160174.1 | - | 1, 2 | 128 | |||
BC117054.1 | - | 1, 2 | 128 | 40126373 | ||
BC117056.1 | - | 1, 2 | 128 | 40126375 | ||
Z21858.1 | - | 1, 2 | 100 |