Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009390.2 | NP_033416.2 | 12 | 2076 | ||
GenBank mRNA | U34042.1 | - | 12 | 2076 |