Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009402.2 | NP_033428.1 | 3 | 543 | ||
GenBank mRNA | AF076482.1 | - | 3 | 542 | ||
AF193843.1 | - | 3 | 531 | |||
AK008335.1 | - | 3 | 544 | |||
BC005582.1 | - | 3 | 553 | 3969014 | ||
CT010165.1 | - | 3 | 507 | |||
X86374.1 | - | 3 | 540 |