Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009409.2 | NP_033435.2 | 18 | 2611 | ||
GenBank mRNA | BC041106.1 | - | 18 | 2631 | 5368508 | |
BC054541.1 | - | 18 | 2611 | 6406462 | ||
D38046.1 | - | 18 | 2184 |