Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009425.2 | NP_033451.1 | 5 | 844 | ||
GenBank mRNA | AK153581.1 | - | 5 | 894 | ||
BC138807.1 | - | 5 | 818 | 8861829 | ||
BC138808.1 | - | 5 | 818 | 8861830 | ||
U37522.1 | - | 5 | 820 |