Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009493.2 | NP_033519.1 | 2 | 423 | ||
GenBank mRNA | AF011414.1 | - | 2 | 423 |
Gene Symbol: Vmn2r32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105063.1 | NP_001098533.1 | 2 | 307 |
Gene Symbol: Vmn2r49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105156.1 | NP_001098626.1 | 2 | 307 |
Set Membership: |
![]() |