Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tuba3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009446.2 | NP_033472.1 | 3 | 350 | ||
GenBank mRNA | AK132775.1 | - | 3 | 350 | ||
BC050770.1 | - | 3 | 336 | 6774775 | ||
BC089547.1 | - | 3 | 336 | 6771690 | ||
M13442.1 | - | 3 | 338 |