Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136086.2 | NP_001129558.1 | 2, 3 | 290 | ||
NM_001291455.1 | NP_001278384.1 | 2, 3 | 540 | |||
NM_009468.5 | NP_033494.1 | 2, 3 | 471 | |||
GenBank mRNA | AF501324.1 | - | 2, 3 | 491 | ||
AK077000.1 | - | 2, 3 | 281 | |||
AK133040.1 | - | 2, 3 | 290 | |||
AK149024.1 | - | 2, 3 | 309 | |||
AK161509.1 | - | 2, 3 | 212 | |||
BC023003.1 | - | 2, 3 | 155 | 5367381 | ||
X87817.1 | - | 2, 3 | 431 |