Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009491.3 | NP_033517.3 | 3 | 874 | ||
GenBank mRNA | AF011426.1 | - | 626 |
Gene Symbol: Vmn2r11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104622.1 | NP_001098092.1 | 3 | 913 |
Gene Symbol: Vmn2r14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104625.1 | NP_001098095.1 | 3 | 874 |
Gene Symbol: Vmn2r15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104626.1 | NP_001098096.1 | 3 | 898 |
Gene Symbol: Vmn2r17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104628.1 | NP_001098098.1 | 3 | 877 |
Gene Symbol: Vmn2r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104620.1 | NP_001098090.1 | 3 | 874 |
Gene Symbol: Vmn2r9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104621.1 | NP_001098091.1 | 3 | 874 |
Set Membership: |
![]() |