Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009518.2 | NP_033544.1 | 4 | 1938 | ||
GenBank mRNA | AK013836.1 | - | 2 | 1325 | ||
BC014737.1 | - | 4 | 1938 | 4921327 | ||
U61969.1 | - | 2448 |