Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009537.3 | NP_033563.2 | 3, 4 | 1014 | ||
GenBank mRNA | AK076149.1 | - | 4, 5 | 627 | ||
AK166542.1 | - | 4, 5 | 352 | |||
BC055899.1 | - | 3, 4 | 992 | 3156776 | ||
M73963.1 | - | 3, 4 | 1014 | |||
M74590.1 | - | 3, 4 | 954 |