Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009556.3 | NP_033582.2 | 4 | 526 | ||
GenBank mRNA | AB221555.1 | - | 1 | 125 | ||
BC053399.1 | - | 4 | 499 | 30031349 | ||
BC098467.1 | - | 4 | 461 | 30008039 | ||
M28382.1 | - | 4 | 487 |