Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291890.1 | NP_001278819.1 | 2, 3 | 131 | ||
NM_001291891.1 | NP_001278820.1 | 2, 3 | 316 | |||
NM_009616.4 | NP_033746.1 | 2, 3 | 316 | |||
GenBank mRNA | AF019887.2 | - | 2, 3 | 221 | ||
AK147217.1 | - | 2, 3 | 239 | |||
AK147549.1 | - | 2, 3 | 132 | |||
BC093480.1 | - | 2, 3 | 307 | |||
CJ111483.1 | - | 2, 3 | 316 | |||
CJ114671.1 | - | 2, 3 | 316 | |||
D50410.2 | - | 2, 3 | 248 |