Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009557.3 | NP_033583.2 | 3 | 2276 | ||
GenBank mRNA | AK051031.1 | - | 3 | 2249 | ||
AK053652.1 | - | 3 | 2281 | |||
AK136405.1 | - | 3 | 2185 | |||
AK148255.1 | - | 3 | 2224 | |||
AK173240.1 | - | 3 | 2262 | |||
BC092398.1 | - | 1 | 1586 | |||
M98502.1 | - | 1 | 1797 |