Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009557.3 | NP_033583.2 | 3 | 3114 | ||
GenBank mRNA | AK136405.1 | - | 3 | 3023 | ||
AK148255.1 | - | 3 | 3065 | |||
AK173240.1 | - | 3 | 3102 | |||
BC092398.1 | - | 1 | 2424 | |||
M98502.1 | - | 1 | 2638 |