Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110794.1 | NP_001104264.1 | 4 | 540 | ||
NM_009674.3 | NP_033804.2 | 4 | 655 | |||
GenBank mRNA | AK032013.1 | - | 4 | 431 | ||
AK077579.1 | - | 4 | 348 | |||
AK134589.1 | - | 4 | 359 | |||
AK149654.1 | - | 4 | 347 | |||
AK151438.1 | - | 4 | 349 | |||
AK161613.1 | - | 4 | 349 | |||
AK167588.1 | - | 4 | 323 | |||
AK169199.1 | - | 4 | 322 | |||
BC008997.1 | - | 4 | 339 | 3157926 | ||
L13129.1 | - | 4 | 342 |