Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF084575.1 | - | 2 | 182 | ||
AK002565.1 | - | 2 | 208 | |||
AK137489.1 | - | 2 | 203 | |||
AK152100.1 | - | 2 | 214 | |||
BC012656.1 | - | 2 | 193 | 4187607 | ||
BC108327.1 | - | 2 | 201 | 30713247 | ||
BC131985.1 | - | 2 | 183 | 40130262 | ||
BC145234.1 | - | 2 | 183 |