Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009718.2 | NP_033848.1 | 2 | 1101 | ||
GenBank mRNA | AK143190.1 | - | 2 | 1109 | ||
BC055743.1 | - | 2 | 1101 | 6808442 | ||
Y07621.1 | - | 2 | 871 |