Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160127.1 | NP_001153599.1 | 2 | 325 | ||
NM_009762.2 | NP_033892.2 | 2 | 325 | |||
GenBank mRNA | AK076266.1 | - | 2 | 289 | ||
AK142153.1 | - | 2 | 293 | |||
AK142252.1 | - | 2 | 303 | |||
BC076601.1 | - | 2 | 303 | 30609878 | ||
BY089666.1 | - | 2 | 326 | |||
U76373.2 | - | 2 | 297 | |||
U76374.2 | - | 2 | 297 |