Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301211.1 | NP_001288140.1 | 4 | 814 | ||
NM_009677.6 | NP_033807.2 | 4 | 814 | |||
GenBank mRNA | AK034198.1 | - | 1 | 43 | ||
AK036387.1 | - | 4 | 757 | |||
AK145821.1 | - | 4 | 736 | |||
BC006691.1 | - | 4 | 491 | |||
BC052703.1 | - | 4 | 687 | 5698075 | ||
BC054535.1 | - | 4 | 687 | 5698075 | ||
X54424.1 | - | 4 | 736 |