Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010261.1 | - | 4 | 320 | ||
AK019319.1 | - | 1 | 56 | |||
AK131624.1 | - | 4 | 320 | |||
AK134921.1 | - | 4 | 320 | |||
AK148747.1 | - | 4 | 321 | |||
AK149111.1 | - | 4 | 322 | |||
AK149568.1 | - | 4 | 302 | |||
AK150834.1 | - | 4 | 322 | |||
AK159105.1 | - | 4 | 320 | |||
AK159424.1 | - | 4 | 322 | |||
AK159517.1 | - | 4 | 320 | |||
BC028816.1 | - | 4 | 299 | 5136415 | ||
BC044785.1 | - | 3 | 291 | |||
BC083351.1 | - | 3 | 289 | 6818357 | ||
CT010212.1 | - | 3 | 254 | |||
CT010356.1 | - | 3 | 254 | |||
DN173921.1 | - | 3 | 422 | |||
M12414.1 | - | 3 | 254 | |||
M73490.1 | - | 2 | 181 |